ENA413171666 Enamine

5-methoxy-N-methylpyrimidin-2-amine

  • MDL number MFCD18816625
  • Cas Number : 31464-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155715

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155715
Availability details for
Quantity
  • Availability
  • Add to Cart