ENA413171668 Enamine

1'-ethyl-1H,1'H-[3,4'-bipyrazole]-4-carboxylic acid

  • MDL number MFCD16109146
  • Cas Number : 1248749-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155718

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155718
Availability details for
Quantity
  • Availability
  • Add to Cart