ENA413171669 Enamine

(2-aminoethyl)trimethylsilane hydrochloride

  • MDL number MFCD12545815
  • Cas Number : 18135-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 300-301
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154925
Availability details for
Quantity
  • Availability
  • Add to Cart