ENA413171671 Enamine

3-(4-bromo-2-chlorophenyl)-1H-pyrazole

  • MDL number MFCD24306029
  • Cas Number : 1554828-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155725

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155725
Availability details for
Quantity
  • Availability
  • Add to Cart