ENA413171672 Enamine

2-chloro-6-(trifluoromethyl)pyridine-3-sulfonyl chloride

  • MDL number MFCD15142816
  • Cas Number : 1208081-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155726

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155726
Availability details for
Quantity
  • Availability
  • Add to Cart