ENA413171676 Enamine

3-amino-1-cyclohexylpiperidin-2-one

  • MDL number MFCD19679251
  • Cas Number : 1343448-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154930

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154930
Availability details for
Quantity
  • Availability
  • Add to Cart