ENA413171683 Enamine

2-methyl-3-(2-oxo-1,2,3,4-tetrahydroquinolin-1-yl)propanenitrile

  • MDL number MFCD11552215
  • Cas Number : 1099685-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-154952

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154952
Availability details for
Quantity
  • Availability
  • Add to Cart