ENA413171686 Enamine

6-(propan-2-yloxy)pyridazin-3-amine

  • MDL number MFCD20703171
  • Cas Number : 1173150-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155747
Availability details for
Quantity
  • Availability
  • Add to Cart