ENA413171689 Enamine

N-[2-(4-bromophenyl)propan-2-yl]cyclobutanamine

  • MDL number MFCD16108435
  • Cas Number : 1249019-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155751

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155751
Availability details for
Quantity
  • Availability
  • Add to Cart