ENA413171691 Enamine

methyl 2-amino-2-(2,4,6-trimethylphenyl)acetate

  • MDL number MFCD12151711
  • Cas Number : 1218141-42-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-155754

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155754
Availability details for
Quantity
  • Availability
  • Add to Cart