ENA413171706 Enamine

4-(bromomethyl)-2-chloro-1-(propan-2-yloxy)benzene

  • MDL number MFCD16075446
  • Cas Number : 936731-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155819

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-155819
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart