ENA413171707 Enamine

6-[3-(4-chlorophenyl)-1,2,4-oxadiazol-5-yl]hexan-1-amine

  • MDL number MFCD12617523
  • Cas Number : 1042514-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155821

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155821
Availability details for
Quantity
  • Availability
  • Add to Cart