ENA413171713 Enamine

6-chloro-9-methyl-9H-purin-2-amine

  • MDL number MFCD13192540
  • Cas Number : 3035-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 268-270
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155833

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155833
Availability details for
Quantity
  • Availability
  • Add to Cart