ENA413171717 Enamine

3-benzoyl-N-(2-chlorophenyl)-1,3-thiazolidine-4-carboxamide

  • MDL number MFCD27500972
  • Cas Number : 1796917-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155199

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155199
Availability details for
Quantity
  • Availability
  • Add to Cart