ENA413171724 Enamine

5-amino-1-methyl-1H-pyrazole-3-carboxylic acid hydrochloride

  • MDL number MFCD25371172
  • Cas Number : 1431966-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155203

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155203
Availability details for
Quantity
  • Availability
  • Add to Cart