ENA413171727 Enamine

2-(4-amino-1H-pyrazol-1-yl)-2-methylpropanoic acid hydrochloride

  • MDL number MFCD25371261
  • Cas Number : 1431963-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155204

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155204
Availability details for
Quantity
  • Availability
  • Add to Cart