ENA413171728 Enamine

furan-2-yl(1H-imidazol-2-yl)methanol

  • MDL number MFCD23866441
  • Cas Number : 1502768-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155888

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155888
Availability details for
Quantity
  • Availability
  • Add to Cart