ENA413171729 Enamine

3-[(3-chlorophenyl)methoxy]-4-methylaniline

  • MDL number MFCD11921151
  • Cas Number : 1153904-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155889

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155889
Availability details for
Quantity
  • Availability
  • Add to Cart