ENA413171739 Enamine

1-[3-(propan-2-yloxy)phenyl]ethan-1-ol

  • MDL number MFCD11520978
  • Cas Number : 1156397-04-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155919

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155919
Availability details for
Quantity
  • Availability
  • Add to Cart