ENA413171741 Enamine

2-(2-aminocyclopentyl)ethan-1-ol

  • MDL number MFCD24448482
  • Cas Number : 109842-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155921

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155921
Availability details for
Quantity
  • Availability
  • Add to Cart