ENA413171743 Enamine

tert-butyl 4-(carbamoylmethyl)piperidine-1-carboxylate

  • MDL number MFCD11046693
  • Cas Number : 782493-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-155940

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155940
Availability details for
Quantity
  • Availability
  • Add to Cart