ENA413171748 Enamine

5-methyl-1-propyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD02855988
  • Cas Number : 890652-02-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-155210

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155210
Availability details for
Quantity
  • Availability
  • Add to Cart