ENA413171750 Enamine

N-(4-chlorophenyl)-4-nitroaniline

  • MDL number MFCD01108458
  • Cas Number : 20983-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155966

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155966
Availability details for
Quantity
  • Availability
  • Add to Cart