ENA413171754 Enamine

5-chloro-1-phenyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD00793866
  • Cas Number : 318288-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155212

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155212
Availability details for
Quantity
  • Availability
  • Add to Cart