ENA413171767 Enamine

6-[1-(hydroxyimino)ethyl]-2H-1,3-benzodioxol-5-amine

  • MDL number MFCD20722539
  • Cas Number : 157593-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156002

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156002
Availability details for
Quantity
  • Availability
  • Add to Cart