ENA413171776 Enamine

1-(4-bromo-2-methylbenzoyl)-4-methylpiperazine

  • MDL number MFCD20543575
  • Cas Number : 924642-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156024

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156024
Availability details for
Quantity
  • Availability
  • Add to Cart