ENA413171784 Enamine

3-(1-ethyl-1H-pyrazol-4-yl)-1-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD16109148
  • Cas Number : 1249995-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156045

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156045
Availability details for
Quantity
  • Availability
  • Add to Cart