ENA413171789 Enamine

5-[(2,3-dihydro-1H-inden-5-yloxy)methyl]furan-2-carbaldehyde

  • MDL number MFCD03422323
  • Cas Number : 832740-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155219
Availability details for
Quantity
  • Availability
  • Add to Cart