ENA413171791 Enamine

2-bromo-2-nitroethan-1-ol

  • MDL number MFCD01728010
  • Cas Number : 5437-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-156055

Safety Information

signalword Danger
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P310,P312,P332+P313,P362,P403+P233,P405,P501
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-156055
pictograms GHS05,GHS07
hstatements H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart