ENA413171793 Enamine

5-[(2,6-dichlorophenoxy)methyl]furan-2-carbaldehyde

  • MDL number MFCD03419438
  • Cas Number : 438218-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155221

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155221
Availability details for
Quantity
  • Availability
  • Add to Cart