ENA413171799 Enamine

1-(cyclopropylmethyl)-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD12804984
  • Cas Number : 1178919-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156075

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-156075
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart