ENA413171801 Enamine

2-(2,3-dihydro-1-benzofuran-5-yl)ethane-1-sulfonyl chloride

  • MDL number MFCD21235688
  • Cas Number : 1523391-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156077

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156077
Availability details for
Quantity
  • Availability
  • Add to Cart