ENA413171807 Enamine

5-chloro-2-[(oxolan-3-yl)methoxy]benzene-1-sulfonyl chloride

  • MDL number MFCD17933571
  • Cas Number : 1285121-04-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156095

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156095
Availability details for
Quantity
  • Availability
  • Add to Cart