ENA413171808 Enamine

2,4,6-triethylbenzaldehyde

  • MDL number MFCD02255729
  • Cas Number : 16688-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155223
Availability details for
Quantity
  • Availability
  • Add to Cart