ENA413171809 Enamine

2-[(2-hydroxy-2-phenylethyl)amino]propan-1-ol

  • MDL number MFCD12771670
  • Cas Number : 42042-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156109

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156109
Availability details for
Quantity
  • Availability
  • Add to Cart