ENA413171812 Enamine

1-{4-[(2-aminophenyl)methyl]piperazin-1-yl}ethan-1-one

  • MDL number MFCD09808106
  • Cas Number : 1016728-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156120

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156120
Availability details for
Quantity
  • Availability
  • Add to Cart