ENA413171818 Enamine

1-(butan-2-yl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD09701732
  • Cas Number : 1170169-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155224

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155224
Availability details for
Quantity
  • Availability
  • Add to Cart