ENA413171829 Enamine

3-(4-methoxy-2-methylphenyl)propanal

  • MDL number MFCD21187722
  • Cas Number : 1477584-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156150

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156150
Availability details for
Quantity
  • Availability
  • Add to Cart