ENA413171838 Enamine

5-chloro-3-(2-methoxyphenyl)-1,2-oxazole

  • MDL number MFCD09749281
  • Cas Number : 1188091-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156198

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156198
Availability details for
Quantity
  • Availability
  • Add to Cart