ENA413171847 Enamine

3-cyclopropyl-1-(2,2,2-trifluoroethyl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD08696564
  • Cas Number : 1006462-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155234
Availability details for
Quantity
  • Availability
  • Add to Cart