ENA413171851 Enamine

2-(2,2-difluoroethoxy)benzaldehyde

  • MDL number MFCD12807550
  • Cas Number : 1184202-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-155237

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155237
Availability details for
Quantity
  • Availability
  • Add to Cart