ENA413171860 Enamine

5,7-dichloro-[1,2,4]triazolo[1,5-a]pyrimidine

  • MDL number MFCD10000845
  • Cas Number : 78706-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131-132
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156302

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156302
Availability details for
Quantity
  • Availability
  • Add to Cart