ENA413171862 Enamine

5-(trifluoromethyl)-1H-pyrazole-3-carbaldehyde

  • MDL number MFCD10697815
  • Cas Number : 591234-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155242

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155242
Availability details for
Quantity
  • Availability
  • Add to Cart