ENA413171863 Enamine

5-nitro-6-(trifluoromethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD01011763
  • Cas Number : 652-43-7
  • Molecular Weight : 225.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-nitro-6-(trifluoromethyl)-2,4-pyrimidinediol
Inchi Code 1S/C5H2F3N3O4/c6-5(7,8)2-1(11(14)15)3(12)10-4(13)9-2/h(H2,9,10,12,13)
Inchi Key BOZXEOUYOAGQOK-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-156332

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156332
Availability details for
Quantity
  • Availability
  • Add to Cart