ENA413171866 Enamine

[3-(1,3-thiazol-2-yl)-1H-pyrazol-5-yl]methanol

  • MDL number MFCD21291264
  • Cas Number : 1798828-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156351

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156351
Availability details for
Quantity
  • Availability
  • Add to Cart