ENA413171867 Enamine

cyclopentyl(4-methylphenyl)methanone

  • MDL number MFCD03841293
  • Cas Number : 97802-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156357

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156357
Availability details for
Quantity
  • Availability
  • Add to Cart