ENA413171871 Enamine

4-methyl-4-(pyridin-3-yl)pyrrolidin-2-one

  • MDL number MFCD20483044
  • Cas Number : 1225218-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156362

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156362
Availability details for
Quantity
  • Availability
  • Add to Cart