ENA413171876 Enamine

dodecyl(2-hydroxyethyl)dimethylazanium bromide

  • MDL number MFCD06252503
  • Cas Number : 7009-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Br-
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-156370

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156370
Availability details for
Quantity
  • Availability
  • Add to Cart