ENA413171880 Enamine

5-[(2,3-dichlorophenoxy)methyl]furan-2-carbaldehyde

  • MDL number MFCD02253767
  • Cas Number : 438219-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155245

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155245
Availability details for
Quantity
  • Availability
  • Add to Cart