ENA413171881 Enamine

2-(2,2-difluoroethoxy)-3-methoxybenzaldehyde

  • MDL number MFCD12768599
  • Cas Number : 1184531-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155246

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155246
Availability details for
Quantity
  • Availability
  • Add to Cart